Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |