Assay Details
| Gene Symbol: MCUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031713.3 | NP_001026883.1 | 9 | 1930 | 89 | |
| GenBank mRNA | BX640678.1 | - | 1 | 324 | 89 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600006937.hg.1 | ENST00000562802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006937.hg.1 | ENST00000562802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006937.hg.1 | ENST00000562802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006937.hg.1 | ENST00000562802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |

MCUR1
Genomic Map