Assay Details
| Gene Symbol: CCDC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011523557.2 | XP_011521859.1 | 1 - 2 | 86 | 79 | |
| GenBank mRNA | AK074059.1 | - | 1 - 2 | 39 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011523557.2 transcripts.
Set Membership: |
|

CCDC57
Genomic Map