Assay Details
| Gene Symbol: CAPN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014289.3 | NP_055104.2 | 13 | 3471 | 145 | |
| GenBank mRNA | AA368247.1 | - | 1 | 145 | ||
| AJ000388.1 | - | 13 | 3492 | 145 | ||
| BC000730.2 | - | 12 | 3300 | 3503916 | 145 | |
| L32067.1 | - | 1 | 145 | |||
More Information
Set Membership: |
|

CAPN6
Genomic Map