Assay Details
| Gene Symbol: FATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033085.2 | NP_149076.1 | 5 | 946 | 74 | |
| GenBank mRNA | AF249872.1 | - | 5 | 946 | 74 | |
| AY101182.1 | - | 5 | 974 | 74 | ||
| BC022064.2 | - | 5 | 947 | 4826440 | 74 | |
More Information
Set Membership: |
|

FATE1
Genomic Map