Assay Details
| Gene Symbol: PRR15L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024320.3 | NP_077296.1 | 2 | 1114 | 68 | |
| XM_005257663.4 | XP_005257720.1 | 2 | 1144 | 68 | ||
| XM_005257664.2 | XP_005257721.1 | 1 | 900 | 68 | ||
| XM_005257665.1 | XP_005257722.1 | 1 | 900 | 68 | ||
| XM_005257666.4 | XP_005257723.1 | 3 | 1225 | 68 | ||
| GenBank mRNA | AA707319.1 | - | 1 | 68 | ||
| AK309589.1 | - | 3 | 665 | 68 | ||
| BC002865.2 | - | 2 | 981 | 3939938 | 68 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AB209036.1 | - | Chr. - - [-] on Build | 68 | |||
More Information
Set Membership: |
|

PRR15L
Genomic Map