Assay Details
| Gene Symbol: FAM71E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145402.1 | NP_001138874.1 | 2 - 3 | 486 | 89 | |
| GenBank mRNA | AK127979.1 | - | 2 - 3 | 433 | 89 | |
| BC031875.1 | - | 2 - 3 | 475 | 89 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |

FAM71E2
Genomic Map