Assay Details
| Gene Symbol: ZNF460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005258422.3 | XP_005258479.1 | 3 | 4497 | 122 | |
| XM_017026176.1 | XP_016881665.1 | 3 | 3947 | 122 | ||
More Information
Set Membership: |
|

ZNF460
Genomic Map