Assay Details
| Gene Symbol: ATP5F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001688.4 | NP_001679.2 | 3 - 4 | 641 | 150 | |
| GenBank mRNA | AK223018.1 | - | 3 - 4 | 266 | 150 | |
| AK291077.1 | - | 3 - 4 | 298 | 150 | ||
| AK308266.1 | - | 4 - 5 | 677 | 150 | ||
| AK313541.1 | - | 3 - 4 | 268 | 150 | ||
| AY871272.1 | - | 3 - 4 | 275 | 150 | ||
| BC005366.1 | - | 3 - 4 | 286 | 3839745 | 150 | |
| BC005960.1 | - | 3 - 4 | 278 | 4049491 | 150 | |
| BC016350.1 | - | 3 - 4 | 317 | 4074170 | 150 | |
| X60221.1 | - | 3 - 4 | 252 | 150 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ATP biosynthetic process
ATP synthesis coupled proton transport
substantia nigra development
mitochondrial ATP synthesis coupled proton transport
protein binding
ATPase activity
transmembrane transporter activity
proton-transporting ATP synthase activity, rotational mechanism
ATP biosynthetic process
ATP synthesis coupled proton transport
substantia nigra development
mitochondrial ATP synthesis coupled proton transport
protein binding
ATPase activity
transmembrane transporter activity
proton-transporting ATP synthase activity, rotational mechanism
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800008964.hg.1 | ENST00000567372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009746.hg.1 | ENST00000397213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009337.hg.1 | ENST00000526403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000464154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000464154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000464154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000464154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000464154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000464154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009746.hg.1 | ENST00000397213.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008964.hg.1 | ENST00000567372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369721.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000369722.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000464154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000464154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000464154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000464154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000464154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000464154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000468818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000483994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | ENST00000493119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009414.hg.1 | NM_001688.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009337.hg.1 | ENST00000526403.1 | Ensembl |

ATP5F1
Genomic Map