Assay Details
| Gene Symbol: HES5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010926.3 | NP_001010926.1 | 3 | 1220 | 74 | |
| XM_005244751.4 | XP_005244808.1 | 2 | 1625 | 74 | ||
| GenBank mRNA | BC087840.1 | - | 1 | 1415 | 6204648 | 74 |
| BX104111.1 | - | 1 | 416 | 74 | ||
| DQ272660.1 | - | 3 | 1220 | 74 | ||
More Information
Set Membership: |
|

HES5
Genomic Map