| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007421.hg.1 |
ENST00000455431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007421.hg.1 |
ENST00000455431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013368.hg.1 |
ENST00000374296.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013368.hg.1 |
ENST00000374296.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013368.hg.1 |
ENST00000374296.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013368.hg.1 |
ENST00000374296.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013368.hg.1 |
NM_178422.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013368.hg.1 |
NM_178422.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013368.hg.1 |
NM_178422.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007421.hg.1 |
ENST00000455431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007421.hg.1 |
ENST00000455431.1 |
Ensembl |