Assay Details
| Gene Symbol: C1orf198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017002599.1 | XP_016858088.1 | 1 - 2 | 531 | 84 | |
| GenBank mRNA | AK131306.1 | - | 1 - 2 | 265 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK131306.1, XM_017002599.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000519360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000519360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000519360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000519360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000519360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000522201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000522201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000522201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000522201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000522201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000522201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000366663.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000427697.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000470540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000519360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000519360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000519360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000519360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000519360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000521263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000522201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000522201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000522201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000522201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000522201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000522201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | ENST00000523410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136494.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_001136495.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_032800.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017708.hg.1 | NM_032800.2 | Refseq |

C1orf198
Genomic Map