Assay Details
| Gene Symbol: SLC41A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK093254.1 | - | 1 | 115 | 94 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |

SLC41A1
Genomic Map