Assay Details
| Gene Symbol: PPM1J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005167.5 | NP_005158.5 | 8 - 9 | 1242 | 94 | |
| GenBank mRNA | AB451327.1 | - | 7 - 8 | 597 | 94 | |
| AB451471.1 | - | 7 - 8 | 597 | 94 | ||
| AK093270.1 | - | 8 - 9 | 1222 | 94 | ||
| AL133644.1 | - | 9 - 10 | 2006 | 94 | ||
| BC011803.2 | - | 8 - 9 | 1166 | 94 | ||
| BC064628.1 | - | 8 - 9 | 1262 | 94 | ||
| BC073828.1 | - | 8 - 9 | 1234 | 5244363 | 94 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |

PPM1J
Genomic Map