Assay Details
| Gene Symbol: SNAP25-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040710.1 | 4 - 5 | 604 | 95 | ||
| GenBank mRNA | AK124961.1 | - | 4 - 5 | 601 | 95 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000438646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000438646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000438646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000438646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000438646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000605592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000605592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000605592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000605592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | ENST00000605592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006683.hg.1 | ENST00000614195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008309.hg.1 | ENST00000460504.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008309.hg.1 | ENST00000460504.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008309.hg.1 | ENST00000625614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012144.hg.1 | ENST00000496127.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012144.hg.1 | ENST00000496127.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012144.hg.1 | ENST00000626470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006683.hg.1 | ENST00000614195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000421143.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000426491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000438646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000438646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000438646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000438646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000438646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000451151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000453544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000603542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000605592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000605592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000605592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000605592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | ENST00000605592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008371.hg.1 | NR_040710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008309.hg.1 | ENST00000460504.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008309.hg.1 | ENST00000460504.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008309.hg.1 | ENST00000625614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012144.hg.1 | ENST00000496127.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012144.hg.1 | ENST00000496127.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012144.hg.1 | ENST00000626470.1 | Ensembl |

SNAP25-AS1
Genomic Map