Assay Details
| Gene Symbol: PPP1R16B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK311244.1 | - | 1 - 2 | 72 | 100 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |

PPP1R16B
Genomic Map