Assay Details
| Gene Symbol: LINC00856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK294518.1 | - | 2 - 3 | 485 | 90 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000421324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000421324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000421324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000421324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000421324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000421324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000432742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000432742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000432742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000432742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000432742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000421324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000421324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000421324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000421324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000421324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000421324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000423770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000432742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000432742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000432742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000432742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000432742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000434974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000510550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | ENST00000624665.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | NR_038985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008164.hg.1 | NR_038985.1 | Refseq |

LINC00856
Genomic Map