Assay Details
| Gene Symbol: PCNX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032223.3 | NP_115599.2 | 23 - 24 | 3810 | 67 | |
| XM_005273987.4 | XP_005274044.1 | 23 - 24 | 5730 | 67 | ||
| XM_006718549.3 | XP_006718612.1 | 23 - 24 | 5715 | 67 | ||
| XM_006718551.3 | XP_006718614.1 | 23 - 24 | 5733 | 67 | ||
| XM_011545024.2 | XP_011543326.1 | 24 - 25 | 4906 | 67 | ||
| XM_011545025.2 | XP_011543327.1 | 23 - 24 | 5733 | 67 | ||
| XM_011545026.2 | XP_011543328.1 | 17 - 18 | 2573 | 67 | ||
| XM_011545027.1 | XP_011543329.1 | 15 - 16 | 1786 | 67 | ||
| GenBank mRNA | BX640978.1 | - | 3 - 4 | 482 | 67 | |
More Information
Set Membership: |
|

PCNX3
Genomic Map