Assay Details
| Gene Symbol: USP41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK303463.1 | - | 1 - 2 | 152 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK303463.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000470202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000470202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000470202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000470202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |

USP41
Genomic Map