Assay Details
| Gene Symbol: OST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134693.1 | NP_001128165.1 | 1 - 2 | 100 | 97 | |
| GenBank mRNA | BC105283.1 | - | 1 - 2 | 58 | 97 | |
| BF697310.1 | - | 1 - 2 | 100 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001134693.1 transcripts.
Set Membership: |
|

OST4
Genomic Map