Assay Details
| Gene Symbol: TCOF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005268508.3 | XP_005268565.1 | 18 - 19 | 3013 | 130 | |
| XM_005268509.3 | XP_005268566.1 | 18 - 19 | 3010 | 130 | ||
More Information
Set Membership: |
|

TCOF1
Genomic Map