Assay Details
| Gene Symbol: MAP3K13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242314.1 | NP_001229243.1 | 6 | 1206 | 85 | |
| NM_001242317.1 | NP_001229246.1 | 4 | 470 | 85 | ||
| NM_004721.4 | NP_004712.1 | 5 | 1321 | 85 | ||
| XM_011513310.2 | XP_011511612.1 | 5 | 1479 | 85 | ||
| XM_017007456.1 | XP_016862945.1 | 6 | 1217 | 85 | ||
| XM_017007457.1 | XP_016862946.1 | 4 | 919 | 85 | ||
| XM_017007458.1 | XP_016862947.1 | 6 | 1107 | 85 | ||
| XM_017007459.1 | XP_016862948.1 | 5 | 1046 | 85 | ||
| GenBank mRNA | AB001872.1 | - | 5 | 1187 | 85 | |
| AK296961.1 | - | 5 | 1249 | 85 | ||
| AK297646.1 | - | 4 | 458 | 85 | ||
| AK302951.1 | - | 5 | 683 | 85 | ||
| AK312714.1 | - | 6 | 1171 | 85 | ||
| BC031677.1 | - | 5 | 939 | 85 | ||
| BC111726.1 | - | 3 | 751 | 40028559 | 85 | |
| Z25428.1 | - | 2 | 228 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111726.1 transcripts.
Set Membership: |
|

MAP3K13
Genomic Map