Assay Details
| Gene Symbol: SORCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020777.2 | NP_065828.2 | 1 | 427 | 79 | |
| XM_005247987.4 | XP_005248044.2 | 1 | 467 | 79 | ||
| XM_011513514.2 | XP_011511816.1 | 1 | 467 | 79 | ||
| XM_011513515.2 | XP_011511817.1 | 1 | 467 | 79 | ||
| XM_011513516.2 | XP_011511818.1 | 1 | 467 | 79 | ||
| XM_017008481.1 | XP_016863970.1 | 1 | 467 | 79 | ||
| GenBank mRNA | AF286190.1 | - | 1 | 326 | 79 | |
| AI206155.1 | - | 1 | 329 | 79 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000505529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000505529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000505529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000505529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000505529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000511199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000511199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000511199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000511199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000511199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | ENST00000511199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000329016.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000505529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000505529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000505529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000505529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000505529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000507866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000511199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000511199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000511199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000511199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000511199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | ENST00000511199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006718.hg.1 | NM_020777.2 | Refseq |

SORCS2
Genomic Map