Assay Details
| Gene Symbol: IPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005270839.4 | XP_005270896.1 | 9 | 4711 | 155 | |
| XM_006710623.3 | XP_006710686.1 | 8 | 4602 | 155 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000430643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000430643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000461718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000461718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009124.hg.1 | ENST00000562232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009124.hg.1 | ENST00000562232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007100.hg.1 | ENST00000546353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007100.hg.1 | ENST00000546353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007100.hg.1 | ENST00000546353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007100.hg.1 | ENST00000546353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000359942.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000396478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000430643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000430643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000461718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000461718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | ENST00000495072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_001145349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014032.hg.1 | NM_005897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007100.hg.1 | ENST00000546353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007100.hg.1 | ENST00000546353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007100.hg.1 | ENST00000546353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007100.hg.1 | ENST00000546353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009124.hg.1 | ENST00000562232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009124.hg.1 | ENST00000562232.1 | Ensembl |

IPP
Genomic Map