Assay Details
| Gene Symbol: SERTAD4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024337.1 | 2 | 611 | 150 | ||
| GenBank mRNA | BC066352.1 | - | 2 | 610 | 150 | |
| BC094794.1 | - | 1 | 2511 | 150 | ||
More Information
Set Membership: |
|

SERTAD4-AS1
Genomic Map