Assay Details
| Gene Symbol: DCAF8L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136533.1 | NP_001130005.1 | 1 | 263 | 86 | |
| GenBank mRNA | BC157859.1 | - | 1 | 263 | 9057078 | 86 |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000578111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000578111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000578111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000578111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000578111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000578111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000580017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000580017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000580017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006856.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000578111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000578111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000578111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000578111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000578111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000578111.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000580017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000580017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000580017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006856.hg.1 | NM_001136533.1 | Refseq |

DCAF8L2
Genomic Map