Assay Details
| Gene Symbol: ZNF268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165881.2 | NP_001159353.1 | 3 | 353 | 114 | |
| NM_001165882.2 | NP_001159354.1 | 3 | 360 | 114 | ||
| NM_001165883.1 | NP_001159355.1 | 3 | 477 | 114 | ||
| NM_001165885.1 | NP_001159357.1 | 3 | 477 | 114 | ||
| NM_003415.2 | NP_003406.1 | 3 | 477 | 114 | ||
| NM_152943.2 | NP_694422.2 | 3 | 477 | 114 | ||
| GenBank mRNA | AF141655.1 | - | 3 | 327 | 114 | |
| AF317549.1 | - | 3 | 477 | 114 | ||
| AF385187.1 | - | 3 | 327 | 114 | ||
| AK027712.1 | - | 3 | 391 | 114 | ||
| AY583691.1 | - | 3 | 308 | 114 | ||
| BC110542.2 | - | 3 | 353 | 40036577 | 114 | |
| CN255799.1 | - | 3 | 337 | 114 | ||
| DQ057356.1 | - | 3 | 266 | 114 | ||
| DQ057358.1 | - | 3 | 266 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF385187.1, AK027712.1, NM_001165882.2 transcripts.
Set Membership: |
|

ZNF268
Genomic Map