Assay Details
| Gene Symbol: TBX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005996.3 | NP_005987.3 | 3 | 1728 | 75 | |
| NM_016569.3 | NP_057653.3 | 4 | 1788 | 75 | ||
| GenBank mRNA | AF002228.1 | - | 3 | 764 | 75 | |
| AF170708.2 | - | 3 | 1024 | 75 | ||
| AF216750.1 | - | 4 | 940 | 75 | ||
| AK054604.1 | - | 3 | 1731 | 75 | ||
| AY034105.1 | - | 2 | 210 | 75 | ||
| BC025258.1 | - | 4 | 1214 | 5458526 | 75 | |
More Information
Set Membership: |
|

TBX3
Genomic Map