| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000554505.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000554505.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000554505.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
ENST00000554505.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000382554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000554505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000554505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000554505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
ENST00000554505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006741.hg.1 |
NM_025081.2 |
Refseq |