Assay Details
| Gene Symbol: LOC115110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037844.2 | 5 | 3762 | 108 | ||
| GenBank mRNA | AK021767.1 | - | 1 | 772 | 108 | |
| AL359943.1 | - | 1 | 2813 | 108 | ||
| BC101995.1 | - | 1 | 264 | 108 | ||
| BC101996.1 | - | 1 | 264 | 108 | ||
| BC101997.1 | - | 1 | 264 | 108 | ||
| BC103697.1 | - | 1 | 264 | 108 | ||
| BC114357.1 | - | 1 | 264 | 108 | ||
| BC114358.2 | - | 1 | 264 | 108 | ||
| CR749717.1 | - | 1 | 2813 | 108 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000416860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000432521.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000443892.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000448624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000448624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000448624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000448624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000448624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000449660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000452793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000452793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000452793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000452793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000452793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | ENST00000452793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012539.hg.1 | NR_037844.2 | Refseq |

LOC115110
Genomic Map