| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008614.hg.1 |
ENST00000377142.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008614.hg.1 |
ENST00000377142.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008614.hg.1 |
ENST00000377142.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008614.hg.1 |
ENST00000377142.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008614.hg.1 |
ENST00000377142.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008614.hg.1 |
NM_002509.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008614.hg.1 |
NM_002509.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008614.hg.1 |
NM_002509.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008614.hg.1 |
NM_002509.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008614.hg.1 |
NM_002509.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006904.hg.1 |
ENST00000549659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006904.hg.1 |
ENST00000549659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008614.hg.1 |
ENST00000377142.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008614.hg.1 |
ENST00000377142.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008614.hg.1 |
ENST00000377142.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008614.hg.1 |
ENST00000377142.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008614.hg.1 |
ENST00000377142.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008614.hg.1 |
NM_002509.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008614.hg.1 |
NM_002509.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008614.hg.1 |
NM_002509.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008614.hg.1 |
NM_002509.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008614.hg.1 |
NM_002509.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006904.hg.1 |
ENST00000549659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006904.hg.1 |
ENST00000549659.1 |
Ensembl |