Assay Details
| Gene Symbol: ADRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054985.1 | - | 871 | 67 | ||
| CR749213.1 | - | 1 | 2044 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADRM1| Gene Symbol: ADRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054985.1 | - | 871 | 67 | ||
| CR749213.1 | - | 1 | 2044 | 67 | ||
Set Membership: |
|