Assay Details
| Gene Symbol: C21orf62-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024622.1 | 4 | 1113 | 111 | ||
| NR_024623.1 | 4 | 1101 | 111 | |||
| GenBank mRNA | AK096601.1 | - | 4 | 1102 | 111 | |
| BC034794.1 | - | 2 | 827 | 111 | ||
| BC117397.1 | - | 4 | 995 | 111 | ||
| BC117399.1 | - | 4 | 1007 | 111 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382377.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000382378.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000477513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000477513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000477513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000477513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000491756.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000491756.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000491756.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000491756.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000491756.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | ENST00000491756.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382375.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382377.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000382378.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000477513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000477513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000477513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000477513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000491756.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000491756.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000491756.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000491756.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000491756.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | ENST00000491756.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024622.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008492.hg.1 | NR_024623.1 | Refseq |

C21orf62-AS1
Genomic Map