Assay Details
| Gene Symbol: DCAF16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017741.3 | NP_060211.3 | 3 | 653 | 109 | |
| XM_005248169.3 | XP_005248226.1 | 3 | 641 | 109 | ||
| XM_005248171.3 | XP_005248228.1 | 2 | 537 | 109 | ||
| XM_006713966.3 | XP_006714029.1 | 4 | 812 | 109 | ||
| XM_017008327.1 | XP_016863816.1 | 2 | 2106 | 109 | ||
| XM_017008328.1 | XP_016863817.1 | 2 | 2091 | 109 | ||
| GenBank mRNA | AK056116.1 | - | 3 | 653 | 109 | |
| BC050697.1 | - | 2 | 314 | 6044417 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK056116.1, BC050697.1, NM_017741.3, XM_005248169.3, XM_005248171.3, XM_006713966.3, XM_017008327.1, XM_017008328.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000418618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000418618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000418618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000418618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000418618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000418618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000570127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000570127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000570127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000570127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | ENST00000570127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000418618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000418618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000418618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000418618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000418618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000418618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000562339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000563550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000566792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000570127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000570127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000570127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000570127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | ENST00000570127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007641.hg.1 | NR_034112.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |

DCAF16
Genomic Map