Assay Details
| Gene Symbol: CXCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK311367.1 | - | 2 | 1322 | 91 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000510390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000510390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000510390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000510390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000510390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | ENST00000395761.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | ENST00000395761.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | ENST00000395761.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | ENST00000395761.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | ENST00000395761.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | ENST00000509101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | ENST00000509101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | ENST00000509101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | ENST00000509101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | ENST00000509101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | NM_001511.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | NM_001511.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | NM_001511.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | NM_001511.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | NM_001511.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | NM_001511.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | NR_046035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | NR_046035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | NR_046035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | NR_046035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | NR_046035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007840.hg.1 | NR_046035.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000296026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000502974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000510390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000510390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000510390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000510390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000510390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | ENST00000511669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011015.hg.1 | NM_002090.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | ENST00000395761.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | ENST00000395761.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | ENST00000395761.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | ENST00000395761.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | ENST00000395761.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | ENST00000509101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | ENST00000509101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | ENST00000509101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | ENST00000509101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | ENST00000509101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | NM_001511.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | NM_001511.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | NM_001511.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | NM_001511.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | NM_001511.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | NM_001511.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | NR_046035.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | NR_046035.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | NR_046035.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | NR_046035.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | NR_046035.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007840.hg.1 | NR_046035.1 | Refseq |

CXCL3
Genomic Map