| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011085.hg.1 |
ENST00000418613.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011085.hg.1 |
ENST00000418613.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011085.hg.1 |
ENST00000418613.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000455918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000455918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000455918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000455918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000455918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009824.hg.1 |
ENST00000567658.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009824.hg.1 |
ENST00000567658.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011085.hg.1 |
ENST00000418613.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011085.hg.1 |
ENST00000418613.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011085.hg.1 |
ENST00000418613.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009824.hg.1 |
ENST00000567658.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009824.hg.1 |
ENST00000567658.1 |
Ensembl |