Assay Details
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138348.4 | NP_612357.4 | 7 | 1482 | 135 | |
| XM_011514154.2 | XP_011512456.1 | 6 | 5092 | 135 | ||
| XM_017010015.1 | XP_016865504.1 | 6 | 5285 | 135 | ||
| GenBank mRNA | AK092203.1 | - | 3 | 802 | 135 | |
| BC007706.2 | - | 5 | 1055 | 3952677 | 135 | |
| BC015392.1 | - | 7 | 1408 | 4430188 | 135 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
sprouting angiogenesis
regulation of tumor necrosis factor-mediated signaling pathway
negative regulation of NF-kappaB transcription factor activity
innate immune response
negative regulation of inflammatory response
canonical Wnt signaling pathway
nucleotide-binding oligomerization domain containing 2 signaling pathway
protein linear deubiquitination
thiol-dependent ubiquitin-specific protease activity
protein binding
cysteine-type peptidase activity
sprouting angiogenesis
regulation of tumor necrosis factor-mediated signaling pathway
negative regulation of NF-kappaB transcription factor activity
innate immune response
negative regulation of inflammatory response
canonical Wnt signaling pathway
nucleotide-binding oligomerization domain containing 2 signaling pathway
protein linear deubiquitination
thiol-dependent ubiquitin-specific protease activity
protein binding
cysteine-type peptidase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000508678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000508678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000508678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000284274.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000503023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000506417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000507335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000508678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000508678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000508678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | ENST00000514913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006822.hg.1 | NM_138348.5 | Refseq |

OTULIN
Genomic Map