Assay Details
| Gene Symbol: NRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270707.1 | NP_001257636.1 | 4 | 1393 | 56 | |
| NM_001270708.1 | NP_001257637.1 | 3 | 1220 | 56 | ||
| NM_001270709.1 | NP_001257638.1 | 3 | 1198 | 56 | ||
| NM_001270710.1 | NP_001257639.1 | 2 | 1001 | 56 | ||
| NM_007243.2 | NP_009174.1 | 5 | 1572 | 56 | ||
| NR_073065.1 | 4 | 1397 | 56 | |||
| NR_073066.1 | 3 | 1178 | 56 | |||
| NR_073072.1 | 3 | 1041 | 56 | |||
| XM_017010224.1 | XP_016865713.1 | 4 | 1574 | 56 | ||
| XM_017010225.1 | XP_016865714.1 | 5 | 1488 | 56 | ||
| GenBank mRNA | AF143676.1 | - | 4 | 1218 | 56 | |
| AI521967.1 | - | 1 | 56 | |||
| AK075509.1 | - | 5 | 1574 | 56 | ||
| AK298152.1 | - | 3 | 1035 | 56 | ||
| AY358411.1 | - | 4 | 1239 | 56 | ||
| BC004509.2 | - | 2 | 844 | 56 | ||
| BC021260.1 | - | 4 | 1210 | 56 | ||
| BC039865.1 | - | 4 | 1238 | 6043663 | 56 | |
| JQ798186.1 | - | 3 | 1061 | 56 | ||
| JQ798187.1 | - | 2 | 864 | 56 | ||
More Information
Set Membership: |
|

NRM
Genomic Map