Assay Details
| Gene Symbol: GPANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199237.1 | NP_001186166.1 | 3 | 1118 | 170 | |
| NM_001199238.1 | NP_001186167.1 | 3 | 987 | 170 | ||
| NM_001199239.1 | NP_001186168.1 | 3 | 984 | 170 | ||
| NM_001199240.1 | NP_001186169.1 | 3 | 770 | 170 | ||
| NM_033177.3 | NP_149417.1 | 2 | 1094 | 170 | ||
| XM_005249403.3 | XP_005249460.1 | 3 | 1121 | 170 | ||
| XM_006715204.1 | XP_006715267.1 | 3 | 799 | 170 | ||
| XM_011514909.1 | XP_011513211.1 | 3 | 839 | 170 | ||
| XM_011514910.1 | XP_011513212.1 | 3 | 836 | 170 | ||
| GenBank mRNA | AJ420471.1 | - | 2 | 686 | 170 | |
| AK123707.1 | - | 3 | 1118 | 170 | ||
| AK314056.1 | - | 3 | 987 | 170 | ||
| BC008783.1 | - | 2 | 1084 | 3629572 | 170 | |
| BC018875.2 | - | 2 | 1078 | 170 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000445768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000445768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000445768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000445768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000445768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000456540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000456540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000456540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000456540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000458083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000458083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000458083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000458083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000458083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | ENST00000458083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375893.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375895.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375896.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375900.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000375906.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000445768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000445768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000445768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000445768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000445768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000456540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000456540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000456540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000456540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000458083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000458083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000458083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000458083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000458083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | ENST00000458083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199240.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011443.hg.1 | NM_001199240.1 | Refseq |

GPANK1
Genomic Map