Assay Details
Gene Symbol: LOC105378146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_136248.1 | 1 | 360 | 89 | ||
GenBank mRNA | BC044924.2 | - | 1 | 322 | 89 | |
BG771988.1 | - | 1 | 323 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
