Assay Details
| Gene Symbol: LINC00667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC119664.1 | - | 1 | 100 | ||
| BC119665.1 | - | 1 | 219 | 100 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | NR_026849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007957.hg.1 | NR_026849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000458396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000458396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000579933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | ENST00000581170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | NR_026849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007957.hg.1 | NR_026849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000458396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000458396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000577439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000580684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000581471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | ENST00000582363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006542.hg.1 | NR_015389.1 | Refseq |

LINC00667
Genomic Map