Assay Details
| Gene Symbol: RIBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144968.3 | NP_659405.1 | 5 | 1386 | 70 | |
| GenBank mRNA | AK057345.1 | - | 5 | 1352 | 70 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000329209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000375327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000414955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000457095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | ENST00000490702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001031745.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_001267053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007325.hg.1 | NM_144968.3 | Refseq |

RIBC1
Genomic Map