| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008980.hg.1 |
ENST00000553425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008980.hg.1 |
ENST00000553425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008980.hg.1 |
ENST00000553425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008980.hg.1 |
ENST00000553425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008980.hg.1 |
ENST00000556024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008980.hg.1 |
ENST00000556024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006946.hg.1 |
ENST00000554829.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006946.hg.1 |
ENST00000554829.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006946.hg.1 |
ENST00000554829.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006946.hg.1 |
ENST00000554829.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008980.hg.1 |
ENST00000553425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008980.hg.1 |
ENST00000553425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008980.hg.1 |
ENST00000553425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008980.hg.1 |
ENST00000553425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008980.hg.1 |
ENST00000556024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008980.hg.1 |
ENST00000556024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006946.hg.1 |
ENST00000554829.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006946.hg.1 |
ENST00000554829.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006946.hg.1 |
ENST00000554829.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006946.hg.1 |
ENST00000554829.1 |
Ensembl |