Assay Details
| Gene Symbol: KTN1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027123.1 | 3 | 1351 | 114 | ||
| GenBank mRNA | BX248296.1 | - | 3 | 1333 | 114 | |
| JX046911.1 | - | 3 | 1356 | 114 | ||
| JX628584.1 | - | 3 | 1356 | 114 | ||
More Information
Set Membership: |
|

KTN1-AS1
Genomic Map