Assay Details
| Gene Symbol: GABPB1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024490.1 | 1 | 2518 | 89 | ||
| GenBank mRNA | BX537481.1 | - | 2 | 671 | 89 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000558593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000558593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000558593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000560359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000560359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000560359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000560359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000561289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000561289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000561289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000583750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000619314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000619314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000619314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000619314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000619314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | ENST00000619314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007199.hg.1 | NR_039862.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000499624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000558593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000558593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000558593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000560359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000560359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000560359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000560359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000561289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000561289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000561289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000583750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000619314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000619314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000619314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000619314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000619314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | ENST00000619314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_024490.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007199.hg.1 | NR_039862.1 | Refseq |

GABPB1-AS1
Genomic Map