| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
ENST00000558623.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
ENST00000558623.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
ENST00000559721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
ENST00000559721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
ENST00000559721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009099.hg.1 |
ENST00000558421.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009099.hg.1 |
ENST00000558421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009099.hg.1 |
ENST00000558421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009099.hg.1 |
ENST00000558421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
ENST00000558623.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
ENST00000558623.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
ENST00000559721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
ENST00000559721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
ENST00000559721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |