Assay Details
| Gene Symbol: ZC3H12D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207360.2 | NP_997243.2 | 6 | 3774 | 113 | |
| GenBank mRNA | AK090441.1 | - | 1 | 4423 | 113 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000409948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000458251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000458251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000458251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000462655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000462655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000462655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000462655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000462655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000498662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | ENST00000498662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000409948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000458251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000458251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000458251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000462655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000462655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000462655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000462655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000462655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000498662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | ENST00000498662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | NM_207360.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013519.hg.1 | NM_207360.2 | Refseq |

ZC3H12D
Genomic Map