Assay Details
| Gene Symbol: MSLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177355.2 | NP_001170826.1 | 14 | 1413 | 70 | |
| NM_005823.5 | NP_005814.2 | 14 | 1473 | 70 | ||
| NM_013404.4 | NP_037536.2 | 13 | 1440 | 70 | ||
| XM_005255034.4 | XP_005255091.1 | 13 | 1719 | 70 | ||
| XM_011522348.2 | XP_011520650.1 | 14 | 1510 | 70 | ||
| XM_017022853.1 | XP_016878342.1 | 13 | 1657 | 70 | ||
| XM_017022854.1 | XP_016878343.1 | 13 | 1668 | 70 | ||
| XM_017022855.1 | XP_016878344.1 | 13 | 1566 | 70 | ||
| XM_017022856.1 | XP_016878345.1 | 13 | 1567 | 70 | ||
| XM_017022857.1 | XP_016878346.1 | 15 | 1587 | 70 | ||
| XM_017022858.1 | XP_016878347.1 | 14 | 1507 | 70 | ||
| GenBank mRNA | AF180951.1 | - | 3 | 417 | 70 | |
| AY743922.1 | - | 14 | 1473 | 70 | ||
| BC003512.1 | - | 13 | 1719 | 3611296 | 70 | |
| BC009272.2 | - | 14 | 1412 | 3957372 | 70 | |
| CB266931.1 | - | 3 | 361 | 70 | ||
| D49441.1 | - | 13 | 1397 | 70 | ||
| U40434.1 | - | 13 | 1422 | 70 | ||
More Information
Set Membership: |
|

MSLN
Genomic Map