Assay Details
| Gene Symbol: LOC105371352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK094336.1 | - | 1 | 218 | 87 | |
| AK125176.1 | - | 1 | 2015 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105371352| Gene Symbol: LOC105371352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK094336.1 | - | 1 | 218 | 87 | |
| AK125176.1 | - | 1 | 2015 | 87 | ||
Set Membership: |
|