| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000377648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
ENST00000495385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006698.hg.1 |
NM_153812.2 |
Refseq |